Order Entry
Poland
ContactUsLinkComponent
Human PK-R1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PK-R1 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3925-96
Supplier:  ANTIBODIES.COM
New Product
Human PK-R1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PK-R1 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3925-96
Supplier:  ANTIBODIES.COM