Order Entry
Poland
ContactUsLinkComponent
Human Entactin/NID ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Entactin/NID in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5855-96
Supplier:  ANTIBODIES.COM
New Product
Human Entactin/NID ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Entactin/NID in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5855-96
Supplier:  ANTIBODIES.COM