Order Entry
Poland
ContactUsLinkComponent
Mouse Pancreatic Polypeptide ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantit ative determination of mouse Pancreatic Polypeptide in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4780-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Pancreatic Polypeptide ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantit ative determination of mouse Pancreatic Polypeptide in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4780-96
Supplier:  ANTIBODIES.COM