Order Entry
Poland
ContactUsLinkComponent
Human Slit3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Slit3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA5557-96
New Product
Human Slit3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Slit3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA5557-96