Order Entry
Poland
ContactUsLinkComponent
Mouse Carbonic Anhydrase 9/CA9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of mouse Carbonic Anhydrase 9/CA9 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5074-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Carbonic Anhydrase 9/CA9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of mouse Carbonic Anhydrase 9/CA9 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5074-96
Supplier:  ANTIBODIES.COM