Multiscreen® SEQ₃₈₄ Filter Plates
Multiscreen® SEQ₃₈₄ filter plate, Clear, 100 µl, 384, -
Catalog # MILYS384SEQ50
New Product
Supplier: MILLIPORE
Jump to:
Product Details & Documents
The MultiScreen384-SEQ filter plate is suited for use with most liquid handling systems. The filter plate operates by vacuum filtration enabling top-loading and recovery of sample. Our plate is designed with the needs of the high throughput laboratory in mind, meets SBS standards and is ideally suited for 'walk away' automation.
- Automation friendly: Design and protocol saves time, money and improves reproducibility
- Efficient dye terminator & salt removal: Recovers highly purified sequencing products for long, accurate sequencing reads
- Eliminates dry-down step: Sequencing products may be resuspended in any aqueous electrokinetic injection buffer
- Saves reagents: plates suitable for use with ¹/₁₆ sequencing reactions
- Proprietary membrane
- Color code: Clear
- Automation compatible: No
Application: Research category, assays, general filtration and nucleic acid.
Packaging: Does not include reagents.
Specifications
- No. of wells:384
- Recommended working volume:100 µl
- Sterile:-
- Colour:Clear
- Description:Multiscreen® SEQ₃₈₄ filter plate



