Order Entry
Poland
ContactUsLinkComponent
Human MSRA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human MSRA in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7743-96
Supplier:  Antibodies.com
New Product
Human MSRA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human MSRA in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7743-96
Supplier:  Antibodies.com