Order Entry
Poland
ContactUsLinkComponent
Mouse MAT1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse MAT1A in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA6246-48
New Product
Mouse MAT1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse MAT1A in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA6246-48