Order Entry
Poland
ContactUsLinkComponent
Human Gankyrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Gankyrin in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6855-48
Supplier:  ANTIBODIES.COM
New Product
Human Gankyrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Gankyrin in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6855-48
Supplier:  ANTIBODIES.COM