Order Entry
Poland
ContactUsLinkComponent
Mouse IL-11RA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse IL-11RA in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA7347-48
New Product
Mouse IL-11RA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse IL-11RA in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA7347-48