Order Entry
Poland
ContactUsLinkComponent
Mouse ZNF179/BFP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse ZNF179/BFP in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7059-96
Supplier:  ANTIBODIES.COM
New Product
Mouse ZNF179/BFP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse ZNF179/BFP in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7059-96
Supplier:  ANTIBODIES.COM