Order Entry
Poland
ContactUsLinkComponent
Human LRRFIP1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human LRRFIP1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7212-48
Supplier:  ANTIBODIES.COM
New Product
Human LRRFIP1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human LRRFIP1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7212-48
Supplier:  ANTIBODIES.COM