Order Entry
Poland
ContactUsLinkComponent
Human NXN/NRX ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human NXN/NRX in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6150-48
Supplier:  Antibodies.com
New Product
Human NXN/NRX ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human NXN/NRX in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6150-48
Supplier:  Antibodies.com