Order Entry
Poland
ContactUsLinkComponent
Mouse MRP6/ARA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse MRP6/ARA in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6937-48
Supplier:  Antibodies.com
New Product
Mouse MRP6/ARA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse MRP6/ARA in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6937-48
Supplier:  Antibodies.com