Order Entry
Poland
ContactUsLinkComponent
Rat Factor XIIIa ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of rat Factor XIIIa in plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6898-48
Supplier:  Antibodies.com
New Product
Rat Factor XIIIa ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of rat Factor XIIIa in plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6898-48
Supplier:  Antibodies.com