Order Entry
Poland
ContactUsLinkComponent
Mouse Ogg1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse Ogg1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7873-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Ogg1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse Ogg1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7873-96
Supplier:  ANTIBODIES.COM