Order Entry
Poland
ContactUsLinkComponent
Human Aurora B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Aurora B in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7773-48
New Product
Human Aurora B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human Aurora B in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7773-48