Order Entry
Poland
ContactUsLinkComponent
Mouse DDT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse DDT in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6424-48
Supplier:  Antibodies.com
New Product
Mouse DDT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse DDT in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6424-48
Supplier:  Antibodies.com