Order Entry
Poland
ContactUsLinkComponent
Mouse Ionotropic Glutamate Receptor 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of mouse Ionotropic Glutamate Receptor 2 in tissue homogenates, cell lysates or other biolo 1 * 48 Tests
Catalog # ANTIA7274-48
Supplier:  Antibodies.com
New Product
Mouse Ionotropic Glutamate Receptor 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro q uantitative determination of mouse Ionotropic Glutamate Receptor 2 in tissue homogenates, cell lysates or other biolo 1 * 48 Tests
Catalog # ANTIA7274-48
Supplier:  Antibodies.com