Order Entry
Poland
ContactUsLinkComponent
Human Talin 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human Talin 2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA6747-48
New Product
Human Talin 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human Talin 2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA6747-48