Order Entry
Poland
ContactUsLinkComponent
Rat CD11a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CD11a in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7137-48
Supplier:  Antibodies.com
New Product
Rat CD11a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CD11a in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7137-48
Supplier:  Antibodies.com