Order Entry
Poland
ContactUsLinkComponent
Human CX3CR1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CX3CR1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7615-48
New Product
Human CX3CR1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CX3CR1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7615-48