Human Cytochrome P450 1A2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cytochrome P450 1A2 in tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA6284-48
Supplier: Antibodies.com