Order Entry
Poland
ContactUsLinkComponent
Human MTCO2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MTCO2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7003-48
New Product
Human MTCO2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MTCO2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7003-48