Order Entry
Poland
ContactUsLinkComponent
Human Cytokeratin 14 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Cytokeratin 14 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA6331-48
New Product
Human Cytokeratin 14 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Cytokeratin 14 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA6331-48