Order Entry
Poland
ContactUsLinkComponent
Human DPAGT1/GPT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human DPAGT1/GPT in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7294-48
Supplier:  ANTIBODIES.COM
New Product
Human DPAGT1/GPT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human DPAGT1/GPT in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7294-48
Supplier:  ANTIBODIES.COM