Order Entry
Poland
ContactUsLinkComponent
Human FLCN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human FLCN in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6462-96
Supplier:  ANTIBODIES.COM
New Product
Human FLCN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human FLCN in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6462-96
Supplier:  ANTIBODIES.COM