Order Entry
Poland
ContactUsLinkComponent
Human ADNP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human ADNP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6031-48
Supplier:  Antibodies.com
New Product
Human ADNP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human ADNP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6031-48
Supplier:  Antibodies.com