Order Entry
Poland
ContactUsLinkComponent
Human NELL2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human NELL2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA6330-48
New Product
Human NELL2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human NELL2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA6330-48