Order Entry
Poland
ContactUsLinkComponent
Rat PTEN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat PTEN in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6980-48
Supplier:  ANTIBODIES.COM
New Product
Rat PTEN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat PTEN in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6980-48
Supplier:  ANTIBODIES.COM