Order Entry
Poland
ContactUsLinkComponent
Rat Claudin 5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat Claudin 5 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6391-48
Supplier:  Antibodies.com
New Product
Rat Claudin 5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat Claudin 5 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6391-48
Supplier:  Antibodies.com