Order Entry
Poland
ContactUsLinkComponent
Human Uroguanylin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Uroguanylin in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6226-48
Supplier:  ANTIBODIES.COM
New Product
Human Uroguanylin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Uroguanylin in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6226-48
Supplier:  ANTIBODIES.COM