Order Entry
Poland
ContactUsLinkComponent
Human MAP1LC3A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human MAP1LC3A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7309-48
Supplier:  Antibodies.com
New Product
Human MAP1LC3A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human MAP1LC3A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7309-48
Supplier:  Antibodies.com