Order Entry
Poland
ContactUsLinkComponent
Mouse Utrophin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse Utrophin in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7021-48
Supplier:  Antibodies.com
New Product
Mouse Utrophin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse Utrophin in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7021-48
Supplier:  Antibodies.com