Order Entry
Poland
ContactUsLinkComponent
Rat PPM1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat PPM1A in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7016-96
Supplier:  ANTIBODIES.COM
New Product
Rat PPM1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat PPM1A in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7016-96
Supplier:  ANTIBODIES.COM