Order Entry
Poland
ContactUsLinkComponent
Human Interferon alpha 10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Interferon alpha 10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA7577-48
Supplier:  Antibodies.com
New Product
Human Interferon alpha 10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Interferon alpha 10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA7577-48
Supplier:  Antibodies.com