Order Entry
Poland
ContactUsLinkComponent
Mouse SLC27A4/FATP4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse SLC27A4/FATP4 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA82374-48
Supplier:  ANTIBODIES.COM
New Product
Mouse SLC27A4/FATP4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse SLC27A4/FATP4 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA82374-48
Supplier:  ANTIBODIES.COM