Order Entry
Poland
ContactUsLinkComponent
Human KDM4A/JHDM3A/JMJD2A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KDM4A/JHDM3A/JMJD2A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7364-48
Supplier:  ANTIBODIES.COM
New Product
Human KDM4A/JHDM3A/JMJD2A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KDM4A/JHDM3A/JMJD2A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7364-48
Supplier:  ANTIBODIES.COM