Order Entry
Poland
ContactUsLinkComponent
Rat CART ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CART in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA7141-48
New Product
Rat CART ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat CART in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA7141-48