Rat DLL4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat DLL4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6441-48
Supplier: Antibodies.com