Order Entry
Poland
ContactUsLinkComponent
Human Kynurenine 3-Monooxygenase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Kynurenine 3-Monooxygenase in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6764-48
Supplier:  ANTIBODIES.COM
New Product
Human Kynurenine 3-Monooxygenase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Kynurenine 3-Monooxygenase in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6764-48
Supplier:  ANTIBODIES.COM