Order Entry
Poland
ContactUsLinkComponent
Mouse FAM135B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse FAM135B in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5968-96
Supplier:  ANTIBODIES.COM
New Product
Mouse FAM135B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of mouse FAM135B in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5968-96
Supplier:  ANTIBODIES.COM