Order Entry
Poland
ContactUsLinkComponent
Human Adenine Nucleotide Translocator 2/ANT 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Adenine Nucleotide Translocator 2/ANT 2 in serum, plasma, tissue homogenat 1 * 96 Tests
Catalog # ANTIA75003-96
Supplier:  ANTIBODIES.COM
New Product
Human Adenine Nucleotide Translocator 2/ANT 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Adenine Nucleotide Translocator 2/ANT 2 in serum, plasma, tissue homogenat 1 * 96 Tests
Catalog # ANTIA75003-96
Supplier:  ANTIBODIES.COM