Order Entry
Poland
ContactUsLinkComponent
Human Parathyroid Hormone Receptor 1/PTH1R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vi tro quantitative determination of human Parathyroid Hormone Receptor 1/PTH1R in tissue homogenates, cell lysates or o 1 * 48 Tests
Catalog # ANTIA7250-48
Supplier:  ANTIBODIES.COM
New Product
Human Parathyroid Hormone Receptor 1/PTH1R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vi tro quantitative determination of human Parathyroid Hormone Receptor 1/PTH1R in tissue homogenates, cell lysates or o 1 * 48 Tests
Catalog # ANTIA7250-48
Supplier:  ANTIBODIES.COM