Order Entry
Poland
ContactUsLinkComponent
Horse TIMP1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of horse TIMP1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6020-48
Supplier:  Antibodies.com
New Product
Horse TIMP1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of horse TIMP1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6020-48
Supplier:  Antibodies.com