Order Entry
Poland
ContactUsLinkComponent
Human ATG3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human ATG3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA82381-48
New Product
Human ATG3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human ATG3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA82381-48