Order Entry
Poland
ContactUsLinkComponent
Mouse Myogenin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse Myogenin in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA6884-48
New Product
Mouse Myogenin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse Myogenin in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA6884-48