Order Entry
Poland
ContactUsLinkComponent
Glucocorticoid ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determin ation of Glucocorticoid in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologic 1 * 48 Tests
Catalog # ANTIA82420-48
Supplier:  ANTIBODIES.COM
New Product
Glucocorticoid ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determin ation of Glucocorticoid in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologic 1 * 48 Tests
Catalog # ANTIA82420-48
Supplier:  ANTIBODIES.COM