Order Entry
Poland
ContactUsLinkComponent
Human EAAT4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human EAAT4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7368-48
Supplier:  ANTIBODIES.COM
New Product
Human EAAT4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human EAAT4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7368-48
Supplier:  ANTIBODIES.COM