Order Entry
Poland
ContactUsLinkComponent
Human Tyrosinase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Tyrosinase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6600-48
Supplier:  Antibodies.com
New Product
Human Tyrosinase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Tyrosinase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6600-48
Supplier:  Antibodies.com